Skip to Content
Merck
All Photos(1)

Documents

15972

Supelco

2,5-Hexanedione

analytical standard

Synonym(s):

Acetonylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
506525
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)
n20/D 1.425

bp

191 °C (lit.)

mp

−6-−5 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(=O)CCC(C)=O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Hexanedione (2,5HD) is a major metabolite of methyl n-butyl ketone. It is neurotoxic in nature and is water soluble.

Application

It was used as reference standard for the determination of 2,5HD in human urine using gas chromatography-electron capture detection and gas chromatography-mass selective detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Spencer et al.
Journal of neurology, neurosurgery, and psychiatry, 38(8), 771-775 (1975-08-01)
Chronic exposure of rats to 2,5-hexanedione (CH3COCH2CH2COCH3), a major metabolite of the neurotoxic industrial solvent methyl n-buryl ketone (CH3COCH2CH2CH2CH3), has been shown to cause a clinical peripheral neuropathy with dying-back peripheral and central nervous system degeneration characterized by giant axonal
Gas chromatographic method for the sensitive determination of 2, 5-hexanedione using electron capture and mass-selective detection.
Konidari, C. N., C. D. Stalikas, and M. I. Karayannis
Analytica Chimica Acta, 442.2, 231-239 (2001)
Hideki Yamasaki et al.
Toxicological sciences : an official journal of the Society of Toxicology, 117(2), 449-456 (2010-07-10)
In the testis, developing germ cells are dependent on supportive physical and paracrine interactions with Sertoli cells. The intimate nature of this relationship is demonstrated by the fact that a toxic insult compromising the stability of Sertoli cells will have
Min-Sun Kim et al.
Toxicology, 260(1-3), 97-103 (2009-05-26)
2,5-Hexanedione (HD), a metabolite of n-hexane, causes central and peripheral neuropathy leading to motor neuron deficits. Although chronic exposure to n-hexane is known to cause gradual sensorimotor neuropathy, there are no reports on the effects of low doses of HD
Yan Sun et al.
Archives of toxicology, 86(2), 205-215 (2011-09-09)
Studies have shown that 2,5-hexanedione (2,5-HD) is the main active metabolite of n-hexane in the human body. The toxicity of n-hexane and 2,5-hexanedione has been extensively researched, but toxicity to the reproductive system, especially the impact on female reproductive function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service