Merck
  • Looping-out mechanism for resolution of replicative stress at telomeres.
Looping-out mechanism for resolution of replicative stress at telomeres.
EMBO reports (2017-06-16)
Tianpeng Zhang, Zepeng Zhang, Feng Li, Qian Hu, Haiying Liu, Mengfan Tang, Wenbin Ma, Junjiu Huang, Zhou Songyang, Yikang Rong, Shichuan Zhang, Benjamin Pc Chen, Yong Zhao
ABSTRACT
Repetitive DNA is prone to replication fork stalling, which can lead to genome instability. Here, we find that replication fork stalling at telomeres leads to the formation of
Sigma-Aldrich
Anti-TRF2 Antibody, clone 4A794, clone 4A794, Upstate®, from mouse