Merck
  • Epilepsy and intellectual disability linked protein Shrm4 interaction with GABA
Epilepsy and intellectual disability linked protein Shrm4 interaction with GABA
Nature communications (2017-03-07)
Jonathan Zapata, Edoardo Moretto, Saad Hannan, Luca Murru, Anna Longatti, Davide Mazza, Lorena Benedetti, Matteo Fossati, Christopher Heise, Luisa Ponzoni, Pamela Valnegri, Daniela Braida, Mariaelvina Sala, Maura Francolini, Jeffrey Hildebrand, Vera Kalscheuer, Francesca Fanelli, Carlo Sala, Bernhard Bettler, Silvia Bassani, Trevor G Smart, Maria Passafaro
ABSTRACT
Shrm4, a protein expressed only in polarized tissues, is encoded by the KIAA1202 gene, whose mutations have been linked to epilepsy and intellectual disability. However, a physiological role for Shrm4 in the brain is yet to be established. Here, we report that Shrm4 is localized to synapses where it regulates dendritic spine morphology and interacts with the C terminus of GABA
Sigma-Aldrich
Kynurenic acid, ≥98%