Merck
  • Synthesis, characterization, and ligand behaviour of a new ditelluroether (C
Synthesis, characterization, and ligand behaviour of a new ditelluroether (C
Dalton transactions (Cambridge, England : 2003) (2016-11-02)
Merja J Poropudas, J Mikko Rautiainen, Raija Oilunkaniemi, Risto S Laitinen
ABSTRACT
The reaction of 1-naphthyl bromide with n-butyl lithium, elemental tellurium, and 1,4-dibromobutane in THF affords both (C
Sigma-Aldrich
1,4-Dibromobutane, 99%