Merck
  • Characterization of an amyloid precursor protein-binding protein Fe65L2 and its novel isoforms lacking phosphotyrosine-interaction domains.
Characterization of an amyloid precursor protein-binding protein Fe65L2 and its novel isoforms lacking phosphotyrosine-interaction domains.
The Biochemical journal (2002-08-03)
Hiroshi Tanahashi, Takeshi Tabira
ABSTRACT
Human Fe65L2 is a member of the Fe65 protein family, which interacts with amyloid precursor protein (APP). Fe65L2 contains an N-terminal WW (Trp-Trp) domain followed by two phosphotyrosine-interaction domains, and consists of 486 amino acids. In the present study, we cloned and characterized two novel isoforms of Fe65L2, designated I-214 and I-245, which are produced by alternative splicing of the RNA. The splicing events disrupt the ability to bind with APP and low-density-lipoprotein-receptor-related protein. Fe65L2 was highly expressed in the brain, whereas I-214 and I-245 were expressed in various tissues. In HEK-293 cells, Fe65L2 was expressed in the nucleus and cytosol, whereas I-245 and I-214 were localized exclusively to the nucleus. The ratio of I-214 to Fe65L2 mRNA was increased by apoptotic stimuli. Although the overexpression of either Fe65L2 or I-214 did not significantly affect the half-life and maturation of APP, or the secretion of secreted APP, the secretion of beta-amyloid peptide (Abeta)40 and Abeta42 was increased by overexpression of Fe65L2, but not by that of I-214. These results suggest that Fe65L2 affects Abeta production and a possible regulation of Fe65L2 function by alternative splicing.