Merck
  • G-protein-coupled receptor GPR161 is overexpressed in breast cancer and is a promoter of cell proliferation and invasion.
G-protein-coupled receptor GPR161 is overexpressed in breast cancer and is a promoter of cell proliferation and invasion.
Proceedings of the National Academy of Sciences of the United States of America (2014-03-07)
Michael E Feigin, Bin Xue, Molly C Hammell, Senthil K Muthuswamy
ABSTRACT
Triple-negative breast cancer (TNBC) accounts for 20% of breast cancer in women and lacks an effective targeted therapy. Therefore, finding common vulnerabilities in these tumors represents an opportunity for more effective treatment. Despite the growing appreciation of G-protein-coupled receptor (GPCR)-mediated signaling in cancer pathogenesis, very little is known about the role GPCRs play in TNBC. Using genomic information of human breast cancer, we have discovered that the orphan GPCR, G-protein-coupled receptor 161 (GPR161) is overexpressed specifically in TNBC and correlates with poor prognosis. Knockdown of GPR161 impairs proliferation of human basal breast cancer cell lines. Overexpression of GPR161 in human mammary epithelial cells increases cell proliferation, migration, intracellular accumulation of E-cadherin, and formation of multiacinar structures in 3D culture. GPR161 forms a signaling complex with the scaffold proteins β-arrestin 2 and Ile Gln motif containing GTPase Activating Protein 1, a regulator of mammalian target of rapamycin complex 1 and E-cadherin. Consistently, GPR161 amplified breast tumors and cells overexpressing GPR161 activate mammalian target of rapamycin signaling and decrease Ile Gln motif containing GTPase Activating Protein 1 phosphorylation. Thus, we identify the orphan GPCR, GPR161, as an important regulator and a potential drug target for TNBC.