- The clinical importance of the trimethadione tolerance test as a method for quantitative assessment of hepatic functional reserve in patients with biliary atresia.
The clinical importance of the trimethadione tolerance test as a method for quantitative assessment of hepatic functional reserve in patients with biliary atresia.
The trimethadione (TMO) tolerance test was performed to evaluate its usefulness in the assessment of hepatic functional reserve in patients with biliary atresia. Nineteen patients with biliary atresia after hepatic portoenterostomy (age range: 2 months to 25 years; sex: 6 males and 13 females) were studied. The study was performed in the morning after a 12-h fast. TMO was given orally, at a dose of 4 mg/kg, with 5 mL of 5% glucose 2 h before breakfast. Blood samples (0.5 mL) were collected to determine serum TMO and dimethadione (DMO), a metabolite of TMO, levels 4 h after the administration of TMO. TMO and DMO were measured by a gas-liquid chromatographic method. A higher total bilirubin level (over 1 mg/dL) in patients with jaundice was reflected in the smaller serum DMO/TMO ratio 4 h after the oral administration of TMO. In addition, these patients with total bilirubin levels of 1 mg/dL or less had a significantly lower DMO/TMO ratio than the control group (healthy subjects). The serum DMO/TMO ratio showed a close correlation with the Child-Pugh score, which is used for overall evaluation of severity of cirrhosis and Mayo risk scores for primary biliary cirrhosis in adults (0.856, P < 0.01 and 0.788, P < 0.01, respectively). The TMO tolerance test shows the benefit of performing a relatively early test of dynamic liver function to evaluate hepatic functional reserve in pre- and post-operative biliary atresia patients.