Merck
  • Comparison of the Antiviral Activity of Remdesivir, Chloroquine, and Interferon-β as Single or Dual Agents Against the Human Beta-Coronavirus OC43.
Comparison of the Antiviral Activity of Remdesivir, Chloroquine, and Interferon-β as Single or Dual Agents Against the Human Beta-Coronavirus OC43.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research (2023-01-19)
Brady T Hickerson, Faruk Sheikh, Raymond P Donnelly, Natalia A Ilyushina
ABSTRACT
The human beta-coronavirus strain, OC43, provides a useful model for testing the antiviral activity of various agents. We compared the activity of several antiviral drugs against OC43, including remdesivir, chloroquine, interferon (IFN)-β, IFN-λ1, and IFN-λ4, in two distinct cell types: human colorectal carcinoma cell line (HCT-8 cells) and normal human bronchial epithelial (NHBE) cells. We also tested whether these agents mediate additive, synergistic, or antagonistic activity against OC43 infection when used in combination. When used as single agents, remdesivir exhibited stronger antiviral activity than chloroquine, and IFN-β exhibited stronger activity than IFN-λ1 or IFN-λ4 against OC43 in both HCT-8 and NHBE cells. Anakinra (IL-1 inhibitor) and tocilizumab (IL-6 inhibitor) did not mediate any antiviral activity. The combination of IFN-β plus chloroquine or remdesivir resulted in higher synergy scores and higher expression of IFN-stimulated genes than did IFN-β alone. In contrast, the combination of remdesivir plus chloroquine resulted in an antagonistic interaction in NHBE cells. Our findings indicate that the combined use of IFN-β plus remdesivir or chloroquine induces maximal antiviral activity against human coronavirus strain OC43 in primary human respiratory epithelial cells. Furthermore, our experimental OC43 virus infection model provides an excellent method for evaluating the biological activity of antiviral drugs.
Sigma-Aldrich
Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D, clone 542-7D, Chemicon®, from mouse