Merck
  • A natural product targets BRD4 to inhibit phase separation and gene transcription.
A natural product targets BRD4 to inhibit phase separation and gene transcription.
iScience (2022-01-25)
Cong Wang, Huasong Lu, Xiangzhong Liu, Xiang Gao, Wenjing Tian, Haifeng Chen, Yuhua Xue, Qiang Zhou
ABSTRACT
The BET-bromodomain protein BRD4 uses two bromodomains to target acetyl-histones and other domains to recruit P-TEFb and other transcription factors to stimulate transcription of proto-oncogenes and key cell identity genes. Recent studies show that its ability to form phase-separated condensates that cluster preferentially at the super-enhancer regions of target genes is key for BRD4 to exert its functions. Here, we describe the identification of a natural product called PCG from polygonum cuspidatum Sieb.et Zucc., a traditional Chinese medicinal herb, that directly binds to BRD4. This binding inhibits BRD4 phase separation, turns dynamic BRD4 nuclear condensates into static aggregates, and effectively shuts down transcription of BRD4-dependent genes. Thus, through PCG we have discovered a BET inhibitor that not only selectively targets BRD4 but also works by suppressing phase separation, a mechanism of action that is different from those of the other known BET inhibitors.
Sigma-Aldrich
(+)-JQ1, ≥98% (HPLC)
Pricing and availability is not currently available.