Merck
  • Contrasting roles for BMP-4 and ventromorphins (BMP agonists) in TGFβ-induced lens EMT.
Contrasting roles for BMP-4 and ventromorphins (BMP agonists) in TGFβ-induced lens EMT.
Experimental eye research (2021-03-29)
Daisy Y Shu, Kevin Ng, Tayler F L Wishart, Juanita Chui, Malin Lundmark, Mary Flokis, Frank J Lovicu
ABSTRACT
Transforming growth factor beta (TGFβ) and bone morphogenetic protein (BMP) signaling play opposing roles in epithelial-mesenchymal transition (EMT) of lens epithelial cells, a cellular process integral to the pathogenesis of fibrotic cataract. We previously showed that BMP-7-induced Smad1/5 signaling blocks TGFβ-induced Smad2/3-signaling and EMT in rat lens epithelial cell explants. To further explore the antagonistic role of BMPs on TGFβ-signaling, we tested the capability of BMP-4 or newly described BMP agonists, ventromorphins, in blocking TGFβ-induced lens EMT. Primary rat lens epithelial explants were treated with exogenous TGFβ2 alone, or in combination with BMP-4 or ventromorphins. Treatment with TGFβ2 induced lens epithelial cells to undergo EMT and transdifferentiate into myofibroblastic cells with upregulated α-SMA and nuclear translocation of Smad2/3 immunofluorescence. BMP-4 was able to suppress this EMT without blocking TGFβ2-nuclear translocation of Smad2/3. In contrast, the BMP agonists, ventromorphins, were unable to block TGFβ2-induced EMT, despite a transient and early ability to significantly reduce TGFβ2-induced nuclear translocation of Smad2/3. This intriguing disparity highlights new complexities in the responsiveness of the lens to differing BMP-related signaling. Further research is required to better understand the antagonistic relationship between TGFβ and BMPs in lens EMT leading to cataract.