Merck
  • The telomere protein tankyrase 1 regulates DNA damage responses at telomeres.
The telomere protein tankyrase 1 regulates DNA damage responses at telomeres.
Aging (2010-11-16)
Sandy Chang
ABSTRACT
The proliferative potential of eukaryotic cells is critically dependent upon the maintenance of functional telomeres, the protein-DNA complexes that cap the ends of chromosomes. A paper published in this issue of Aging describes that the telomere protein tankyrase 1 regulates DNA damage responses at telomeres.
Sigma-Aldrich
PARP5A active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥30% (SDS-PAGE)