Merck
  • Mesoporogen-Free Synthesis of Hierarchical SAPO-34 with Low Template Consumption and Excellent Methanol-to-Olefin Conversion.
Mesoporogen-Free Synthesis of Hierarchical SAPO-34 with Low Template Consumption and Excellent Methanol-to-Olefin Conversion.
ChemSusChem (2018-09-05)
Qiming Sun, Ning Wang, Risheng Bai, Guangrui Chen, Zhiqiang Shi, Yongcun Zou, Jihong Yu
ABSTRACT
Significant interest has emerged in the development of nanometer-sized and hierarchical silicoaluminophosphate zeolites (SAPO-34) because of their enhanced accessibility and improved catalytic activity for methanol-to-olefin (MTO) conversion. A series of nanometer-sized SAPO-34 catalysts with tunable hierarchical structures was synthesized in a Al2 O3 /H3 PO4 /SiO2 /triethylamine(TEA)/H2 O system by using a mesoporogen-free nanoseed-assisted method. The nanometer-sized hierarchical SH -3.0 catalyst (TEA/Al2 O3 =3.0) possessed the highest crystallinity, highest abundance of intracrystalline meso-/macropores, and the most suitable acidity among all obtained catalysts, showing the highest ethylene and propylene selectivity of 85.4 %. This is the highest reported selectivity for MTO reactions under similar conditions. Detailed analysis of the coke produced during the reaction revealed that the small-sized methyl-substituted benzene and bulky methyl-substituted pyrene were mainly located inside the crystals instead of on the surface of the crystals, which provided further insight into understanding the deactivation of the SAPO-34 catalyst during MTO reaction. Significantly, the simple and cost-effective synthetic process and superb catalytic performance of the nanometer-sized hierarchical SAPO-34 is promising for their practical large-scale application for MTO conversion.