Skip to Content
Merck
EVEROLIMUS SYSTEM SUITABILITY MIXTURE United States Pharmacopeia (USP) Reference Standard

Everolimus

EVEROLIMUS SYSTEM SUITABILITY MIXTURE United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Everolimus, 42-O-(2-Hydroxyethyl)rapamycin
Empirical Formula (Hill Notation):
C53H83NO14
CAS Number:
Molecular Weight:
958.22

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique