Skip to Content
Merck
Clarithromycin for peak identification European Pharmacopoeia (EP) Reference Standard

Clarithromycin

Clarithromycin for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Clarithromycin
Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95

Products

Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique