tert-Butanol
Synonym(s):
2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol, tert-Butanol
Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing