Skip to Content
Merck
Noradrenaline acid tartrate British Pharmacopoeia (BP) Reference Standard

Norepinephrine bitartrate

Noradrenaline acid tartrate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, L-(−)-Noradrenaline (+)-bitartrate salt monohydrate, L-(−)-Norepinephrine-(+)-bitartrate salt monohydrate
Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade