Skip to Content
Merck
N-Nitrosodi-n-butylamine analytical standard

N-Nitrosodi-n-butylamine

N-Nitrosodi-n-butylamine analytical standard
Synonym(s):
N-Nitrosodibutylamine, Dibutyl nitrosamine, NDBA, N-Butyl-N-nitroso-1-butanamine, N-Nitrosodi-n-butylamine solution
Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique