L-Methionine
Synonym(s):
(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid, L-Methionine
Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing