Skip to Content
Merck
vibrant-m

Pholcodine for impurity G identification

vibrant-m
Synonym(s):
Pholcodine
Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50

Products

Formula Weight
API Family
Reference Material Type
Showing 1-1 of 1