α-Cyclodextrin
Synonym(s):
α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose, α-Cyclodextrine, α-Cyclodextrin
Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Sterility
Technique