Skip to Content
Merck
p-Nitroblue Tetrazolium Chloride NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC₅₀ = 3-4 µM).

p-Nitroblue Tetrazolium Chloride

p-Nitroblue Tetrazolium Chloride NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC₅₀ = 3-4 µM).
Synonym(s):
p-Nitroblue Tetrazolium Chloride
Empirical Formula (Hill Notation):
C40H30Cl2N10O6
CAS Number:
Molecular Weight:
817.64

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment