Skip to Content
Merck
Ethanolamine EMPROVE® EXPERT, NF

Ethanolamine

Ethanolamine EMPROVE® EXPERT, NF
Synonym(s):
Ethanolamine, 2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine
Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade