Skip to Content
Merck
Sodium hydroxide puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets

Sodium hydroxide

Sodium hydroxide puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets
Synonym(s):
‘Caustic soda’, Sodium hydroxide, Sodium hydroxide monohydrate
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 22
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2