Progesterone
Synonym(s):
4-Pregnene-3,20-dione, Progesterone
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Sterility
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing