Skip to Content
Merck
Poly(ethylene glycol) dimethyl ether average Mn ~500, contains 100 ppm BHT as stabilizer

Poly(ethylene glycol) dimethyl ether

Poly(ethylene glycol) dimethyl ether average Mn ~500, contains 100 ppm BHT as stabilizer
Synonym(s):
Dimethyl polyethylene glycol, Polyethylene glycol dimethyl ether 500, Polyethylene glycol dimethyl ether 2000, Polyethylene glycol dimethyl ether 250
Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
Molecular Weight:
530.65

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type