Stability and Reproducibility of Zenix® 300 under High Salt and High pH Mobile Phase Conditions
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-150 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steelCONDITIONS
column
Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777018)
mobile phase
50 mM sodium phosphate + 1 M NaCl, pH 8.5
flow rate
1.0 mL/min
pressure
1450 psi (100 bar)
column temp.
25 °C
detector
UV, 214 nm
injection
5 μL
sample
each compound 1 mg/mL except uracil, 0.1 mg/mL
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies