Merck
  • Qualitative differences in T-cell activation by dendritic cell-derived extracellular vesicle subtypes.
Qualitative differences in T-cell activation by dendritic cell-derived extracellular vesicle subtypes.
The EMBO journal (2017-09-20)
Mercedes Tkach, Joanna Kowal, Andres E Zucchetti, Lotte Enserink, Mabel Jouve, Danielle Lankar, Michael Saitakis, Lorena Martin-Jaular, Clotilde Théry
ABSTRACT
Exosomes, nano-sized secreted extracellular vesicles (EVs), are actively studied for their diagnostic and therapeutic potential. In particular, exosomes secreted by dendritic cells (DCs) have been shown to carry MHC-peptide complexes allowing efficient activation of T lymphocytes, thus displaying potential as promoters of adaptive immune responses. DCs also secrete other types of EVs of different size, subcellular origin and protein composition, whose immune capacities have not been yet compared to those of exosomes. Here, we show that large EVs (lEVs) released by human DCs are as efficient as small EVs (sEVs), including exosomes, to induce CD4
Sigma-Aldrich
Anti-CD9 Antibody, clone MM2/57, clone MM2/57, Chemicon®, from mouse
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor