Merck
Fluid shear stress increases transepithelial transport of Ca
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-01-20)
Sami G Mohammed, Francisco J Arjona, Femke Latta, René J M Bindels, Ronald Roepman, Joost G J Hoenderop
ABSTRACT
In kidney, transcellular transport of Ca