Merck
  • The Actinobacillus actinomycetemcomitans autotransporter adhesin Aae exhibits specificity for buccal epithelial cells from humans and old world primates.
The Actinobacillus actinomycetemcomitans autotransporter adhesin Aae exhibits specificity for buccal epithelial cells from humans and old world primates.
Infection and immunity (2005-03-24)
Daniel H Fine, Kabilan Velliyagounder, David Furgang, Jeffrey B Kaplan
ABSTRACT
Cells of the gram-negative periodontopathogen Actinobacillus actinomycetemcomitans express a surface-exposed, outer membrane autotransporter protein, designated Aae, which has been implicated in epithelial cell binding. We constructed a mutant strain of A. actinomycetemcomitans that contained a transposon insertion in the Aae structural gene (aae) and tested the mutant to determine its ability to bind to buccal epithelial cells (BECs) isolated from healthy volunteers. Significantly fewer mutant cells than wild-type cells bound to BECs. A broad-host-range plasmid that contained an intact aae gene driven by a heterologous tac promoter restored the ability of the mutant strain to bind to BECs at wild-type levels. This plasmid also conferred upon Escherichia coli the ability to express the Aae protein on its surface and to bind to human BECs. Aae-expressing E. coli also bound to BECs isolated from six Old World primates but not to BECs isolated from four New World primates or nine other nonprimate mammals, as well as to human gingival epithelial cells but not to human pharyngeal, palatal, tongue, bronchial, or cervical epithelial cells. Our findings indicate that Aae mediates binding of A. actinomycetemcomitans to BECs from humans and Old World primates and that this process may contribute to the host range specificity and tissue tropism exhibited by this bacterium.