- A novel duodenal iron-regulated transporter, IREG1, implicated in the basolateral transfer of iron to the circulation.
A novel duodenal iron-regulated transporter, IREG1, implicated in the basolateral transfer of iron to the circulation.
Iron absorption by the duodenal mucosa is initiated by uptake of ferrous Fe(II) iron across the brush border membrane and culminates in transfer of the metal across the basolateral membrane to the portal vein circulation by an unknown mechanism. We describe here the isolation and characterization of a novel cDNA (Ireg1) encoding a duodenal protein that is localized to the basolateral membrane of polarized epithelial cells. Ireg1 mRNA and protein expression are increased under conditions of increased iron absorption, and the 5' UTR of the Ireg1 mRNA contains a functional iron-responsive element (IRE). IREG1 stimulates iron efflux following expression in Xenopus oocytes. We conclude that IREG1 represents the long-sought duodenal iron export protein and is upregulated in the iron overload disease, hereditary hemochromatosis.