Merck
  • UBQLN2 mutation causing heterogeneous X-linked dominant neurodegeneration.
UBQLN2 mutation causing heterogeneous X-linked dominant neurodegeneration.
Annals of neurology (2014-04-29)
Akl C Fahed, Barbara McDonough, Cynthia M Gouvion, Kathy L Newell, Leon S Dure, Martina Bebin, Alexander G Bick, J G Seidman, Donald H Harter, Christine E Seidman
ABSTRACT
We report a 5-generation family with phenotypically diverse neurodegenerative disease including relentlessly progressive choreoathetoid movements, dysarthria, dysphagia, spastic paralysis, and behavioral dementia in descendants of a 67-year-old woman with amyotrophic lateral sclerosis. Disease onset varied with gender, occurring in male children and adult women. Exome sequence analyses revealed a novel mutation (c.1490C>T, p.P497L) in the ubiquilin-2 gene (UBQLN2) with X-linked inheritance in all studied affected individuals. As ubiquilin-2-positive inclusions were identified in brain, we suggest that mutant peptide predisposes to protein misfolding and accumulation. Our findings expand the spectrum of neurodegenerative phenotypes caused by UBQLN2 mutations.