Merck
  • BODIPY dyes with β-conjugation and their applications for high-efficiency inverted small molecule solar cells.
BODIPY dyes with β-conjugation and their applications for high-efficiency inverted small molecule solar cells.
Chemical communications (Cambridge, England) (2012-07-31)
Hsin-Yu Lin, Wei-Ching Huang, Yung-Chung Chen, Hsien-Hsin Chou, Chih-Yu Hsu, Jiann T Lin, Hao-Wu Lin
ABSTRACT
Small molecule BODIPY dyes incorporating conjugated substituents at the β sites have been synthesized. Solution processed inverted bulk heterojunction cells were fabricated from the blends of the dyes and PC(71)BM. The cells exhibited very high open-circuit voltages (>0.9 V) and a high conversion efficiency of 3.22% has been achieved.
Sigma-Aldrich
Difluoro{2-[1-(3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-3,5-dimethyl-1H-pyrrolato-N}boron, 99% (HPLC)