Merck
  • Primary pulmonary paraganglioma: report of a functioning case with immunohistochemical and ultrastructural study.
Primary pulmonary paraganglioma: report of a functioning case with immunohistochemical and ultrastructural study.
The American journal of surgical pathology (2004-05-29)
Junji Shibahara, Akiteru Goto, Toshiro Niki, Makoto Tanaka, Jun Nakajima, Masashi Fukayama
ABSTRACT
We describe a case of primary pulmonary paraganglioma, a tumor that has not been reported in sufficient detail in previous literature. The patient was a 55-year-old woman with hypertension accompanied by an elevated serum norepinephrine level (2651 pg/mL; normal 100-450 pg/mL). Computed tomography revealed a well-circumscribed solid mass, 3.5 cm in diameter, located in the lower lobe of the left lung. In the lobectomy specimen, the tumor had invaded the B8 bronchus and hilar lymph nodes with microscopic metastasis to the mediastinal nodes. The tumor showed histologic, immunohistochemical, and ultrastructural features of paraganglioma: argyrophilic cells arranged in a nesting (Zellballen) or anastomosing trabecular pattern within an arcuate vascular network. Neoplastic chief cells positive for neuroendocrine markers (CD56, synaptophysin, chromogranin A) were surrounded by sustentacular cells positive for S-100 protein. Neurofilament protein was positively stained, but cytokeratins were totally negative. On electron microscopy, chief cells possessed abundant dense core granules with an eccentric halo ("norepinephrine-type" granules). The patient's blood pressure began to decline soon after the resection, and her serum norepinephrine promptly returned to almost normal. On the basis of our experience, our case is a bona fide primary pulmonary paraganglioma, a tumor heretofore subject to considerable skepticism.
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline