Merck
  • Selective inhibitors of the mutant B-Raf pathway: discovery of a potent and orally bioavailable aminoisoquinoline.
Selective inhibitors of the mutant B-Raf pathway: discovery of a potent and orally bioavailable aminoisoquinoline.
Journal of medicinal chemistry (2009-09-22)
Adrian L Smith, Frenel F DeMorin, Nick A Paras, Qi Huang, Jeffrey K Petkus, Elizabeth M Doherty, Thomas Nixey, Joseph L Kim, Douglas A Whittington, Linda F Epstein, Matthew R Lee, Mark J Rose, Carol Babij, Manory Fernando, Kristen Hess, Quynh Le, Pedro Beltran, Josette Carnahan
ABSTRACT
The discovery and optimization of a novel series of aminoisoquinolines as potent, selective, and efficacious inhibitors of the mutant B-Raf pathway is presented. The N-linked pyridylpyrimidine benzamide 2 was identified as a potent, modestly selective inhibitor of the B-Raf enzyme. Replacement of the benzamide with an aminoisoquinoline core significantly improved kinase selectivity and imparted favorable pharmacokinetic properties, leading to the identification of 1 as a potent antitumor agent in xenograft models.