Merck
  • MAP3K-related kinase involved in NF-kappaB induction by TNF, CD95 and IL-1.
MAP3K-related kinase involved in NF-kappaB induction by TNF, CD95 and IL-1.
Nature (1997-02-06)
N L Malinin, M P Boldin, A V Kovalenko, D Wallach
ABSTRACT
Several members of the tumour-necrosis/nerve-growth factor (TNF/NGF) receptor family activate the transcription factor NF-kappaB through a common adaptor protein, Traf2 (refs 1-5), whereas the interleukin 1 type-I receptor activates NF-kappaB independently of Traf2 (ref. 4). We have now cloned a new protein kinase, NIK, which binds to Traf2 and stimulates NF-kappaB activity. This kinase shares sequence similarity with several MAPKK kinases. Expression in cells of kinase-deficient NIK mutants fails to stimulate NF-kappaB and blocks its induction by TNF, by either of the two TNF receptors or by the receptor CD95 (Fas/Apo-1), and by TRADD, RIP and MORT1/FADD, which are adaptor proteins that bind to these receptors. It also blocked NF-kappaB induction by interleukin-1. Our findings indicate that NIK participates in an NF-kappaB-inducing signalling cascade common to receptors of the TNF/NGF family and to the interleukin-1 type-I receptor.