Merck
  • Phosphodiesterase type 5 inhibitors enhance chemotherapy in preclinical models of esophageal adenocarcinoma by targeting cancer-associated fibroblasts.
Phosphodiesterase type 5 inhibitors enhance chemotherapy in preclinical models of esophageal adenocarcinoma by targeting cancer-associated fibroblasts.
Cell reports. Medicine (2022-06-23)
Benjamin P Sharpe, Annette Hayden, Antigoni Manousopoulou, Andrew Cowie, Robert C Walker, Jack Harrington, Fereshteh Izadi, Stella P Breininger, Jane Gibson, Oliver Pickering, Eleanor Jaynes, Ewan Kyle, John H Saunders, Simon L Parsons, Alison A Ritchie, Philip A Clarke, Pamela Collier, Nigel P Mongan, David O Bates, Kiren Yacqub-Usman, Spiros D Garbis, Zoë Walters, Matthew Rose-Zerilli, Anna M Grabowska, Timothy J Underwood
ABSTRACT
The chemotherapy resistance of esophageal adenocarcinomas (EACs) is underpinned by cancer cell extrinsic mechanisms of the tumor microenvironment (TME). We demonstrate that, by targeting the tumor-promoting functions of the predominant TME cell type, cancer-associated fibroblasts (CAFs) with phosphodiesterase type 5 inhibitors (PDE5i), we can enhance the efficacy of standard-of-care chemotherapy. In ex vivo conditions, PDE5i prevent the transdifferentiation of normal fibroblasts to CAF and abolish the tumor-promoting function of established EAC CAFs. Using shotgun proteomics and single-cell RNA-seq, we reveal PDE5i-specific regulation of pathways related to fibroblast activation and tumor promotion. Finally, we confirm the efficacy of PDE5i in combination with chemotherapy in close-to-patient and in vivo PDX-based model systems. These findings demonstrate that CAFs drive chemotherapy resistance in EACs and can be targeted by repurposing PDE5i, a safe and well-tolerated class of drug administered to millions of patients world-wide to treat erectile dysfunction.
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder