- A truncated protein product of the germline variant of the DUOX2 gene leads to adenomatous polyposis.
A truncated protein product of the germline variant of the DUOX2 gene leads to adenomatous polyposis.
In some patients with adenomatous polyposis, an identifiable pathogenic variant of known associated genes cannot be found. Researchers have studied this for decades; however, few new genes have been identified. Adenomatous polyposis coli (APC) negative polyposis patients were identified through next-generation sequencing and multiplex ligation-dependent probe amplification. Then, whole-exome sequencing (WES) was used to determine candidate genes harboring pathogenic variants. Functional experiments were performed to explore their effects. Subsequently, using Sanger sequencing, we found other polyposis patients carrying variants of the DUOX2 gene, encoding dual oxidase 2, and analyzed them. From 88 patients with suspected familial adenomatous polyposis, 25 unrelated APC negative polyposis patients were identified. Based on the WES results of 3 patients and 2 healthy relatives from a family, the germline nonsense variant (c.1588A>T; p.K530X) of the DUOX2 gene was speculated to play a decisive role in the pedigree in relation to adenomatous polyposis. During functional experiments, we observed that the truncated protein, hDuox2 K530, was overexpressed in the adenoma in a carrier of the DUOX2 nonsense variant, causing abnormal cell proliferation through endoplasmic reticulum (ER) retention. In addition, we found two unrelated APC negative patients carrying DUOX2 missense variants (c.3329G>A, p.R1110Q; c.4027C>T, p.L1343F). Given the results of the in silico analysis, these two missense variants might exert a negative influence on the function of hDuox2. To our knowledge, this is the first study that reports the possible association of DUOX2 germline variants with adenomatous polyposis. With an autosomal dominant inheritance, it causes ER retention, inducing an unfolded protein response.