Merck
  • Conditional deletion of Nedd4-2 in lung epithelial cells causes progressive pulmonary fibrosis in adult mice.
Conditional deletion of Nedd4-2 in lung epithelial cells causes progressive pulmonary fibrosis in adult mice.
Nature communications (2020-04-26)
Julia Duerr, Dominik H W Leitz, Magdalena Szczygiel, Dmytro Dvornikov, Simon G Fraumann, Clemens Kreutz, Piotr K Zadora, Ayça Seyhan Agircan, Philip Konietzke, Theresa A Engelmann, Jan Hegermann, Surafel Mulugeta, Hiroshi Kawabe, Lars Knudsen, Matthias Ochs, Daniela Rotin, Thomas Muley, Michael Kreuter, Felix J F Herth, Mark O Wielpütz, Michael F Beers, Ursula Klingmüller, Marcus A Mall
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a chronic progressive interstitial lung disease characterized by patchy scarring of the distal lung with limited therapeutic options and poor prognosis. Here, we show that conditional deletion of the ubiquitin ligase Nedd4-2 (Nedd4l) in lung epithelial cells in adult mice produces chronic lung disease sharing key features with IPF including progressive fibrosis and bronchiolization with increased expression of Muc5b in peripheral airways, honeycombing and characteristic alterations in the lung proteome. NEDD4-2 is implicated in the regulation of the epithelial Na+ channel critical for proper airway surface hydration and mucus clearance and the regulation of TGFβ signaling, which promotes fibrotic remodeling. Our data support a role of mucociliary dysfunction and aberrant epithelial pro-fibrotic response in the multifactorial disease pathogenesis. Further, treatment with the anti-fibrotic drug pirfenidone reduced pulmonary fibrosis in this model. This model may therefore aid studies of the pathogenesis and therapy of IPF.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Clara Cell Secretory Protein Antibody, serum, Upstate®