- Comparison of Recombinant Proteins from Schistosoma japonicum for Schistosomiasis Diagnosis.
Comparison of Recombinant Proteins from Schistosoma japonicum for Schistosomiasis Diagnosis.
Clinical and vaccine immunology : CVI (2010-01-08)
Ya-mei Jin, Ke Lu, Wei-Fang Zhou, Zhi-Qiang Fu, Jin-Ming Liu, Yao-Jun Shi, Hao Li, Jiao-Jiao Lin
PMID20053872
ABSTRACT
The most important animal reservoirs of Schistosoma japonicum in China are bovines. Diagnosis and control of bovine schistosomiasis is critical for reducing the prevalence of the disease. We screened defined diagnostic antigens that have the potential to increase the sensitivity and specificity of serological assays and to distinguish between active and prior infections. Five recombinant proteins with the potential to be diagnostic antigens were compared to the native soluble egg antigen preparation by enzyme-linked immunosorbent assay (ELISA). We evaluated the potentials of the recombinant proteins for discriminating active from prior infections, as well as the therapeutic efficacy of the established ELISA technique.