Merck
  • Stage-specific expression of the Kit receptor and its ligand (KL) during male gametogenesis in the mouse: a Kit-KL interaction critical for meiosis.
Stage-specific expression of the Kit receptor and its ligand (KL) during male gametogenesis in the mouse: a Kit-KL interaction critical for meiosis.
Development (Cambridge, England) (1998-10-21)
S Vincent, D Segretain, S Nishikawa, S I Nishikawa, J Sage, F Cuzin, M Rassoulzadegan
ABSTRACT
The Kit receptor and its ligand KL, which together constitute an essential effector at various stages of embryonic development, are both present during adult gametogenesis. In the testis, KL is expressed in Sertoli cells, and Kit in germ cells, starting at the premeiotic stages. A series of observations indicated previously a role in spermatogonia survival, without excluding a possible function at later stages. We identified a complex pattern of expression of the two components in the adult murine testis, suggestive of a role in the meiotic progression of spermatocytes. At stages VII-VIII of the cycle of the seminiferous epithelium, the time when spermatocytes enter meiosis, the membrane-associated form of KL extends on the Sertoli cell from the peripheral to the adluminal compartment of the tubule. We also found that the receptor is present on the surface of germ cells up to the pachytene stage. The availability of differentiated Sertoli cell lines, which express the KL protein and support part of the maturation of germ cells in coculture, allowed us to ask whether, in the in vitro reconstructed system, transit of spermatocytes through meiosis requires the Kit-KL interaction. Addition of a blocking monoclonal antibody against the Kit receptor (ACK2) inhibited extensively the appearance of haploid cells and the expression of a haploid-phase-specific gene (Prm1). Recognition of the supporting Sertoli cell by germ cells was not affected, indicating a requirement for the activity of the receptor for either entering or completing meiosis. Involvement of the membrane-associated form of the ligand was suggested by the observation that addition of the soluble form of KL was equally inhibitory.