Skip to Content
Merck
Ketoconazole British Pharmacopoeia (BP) Reference Standard

Ketoconazole

Ketoconazole British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Ketoconazole, (±)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine
Empirical Formula (Hill Notation):
C26H28Cl2N4O4
CAS Number:
Molecular Weight:
531.43
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique