Skip to Content
Merck
Valaciclovir for system suitability European Pharmacopoeia (EP) Reference Standard

Valacyclovir HCl

Valaciclovir for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Valacyclovir hydrochloride
Empirical Formula (Hill Notation):
C13H20N6O4 · HCl
CAS Number:
Molecular Weight:
360.80

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique