Butylparaben
Synonym(s):
4-Hydroxybenzoic acid butyl ester, Butylparaben
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing