Enoxaparin sodium for Bioassays
Synonym(s):
Heparin sodium salt, Heparin sodium, Heparin sodium salt solution
CAS Number:
Products
Brand
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-18 of 18
Compare
Product Number
Description
Pricing