Skip to Content
Merck
Agar Strips SDX Sabouraud Dextrose Agar, Yeasts and molds, HYCON®, sterile; aseptically filled

Agar Strips SDX

Agar Strips SDX Sabouraud Dextrose Agar, Yeasts and molds, HYCON®, sterile; aseptically filled
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Product Line
Compatibility
Quality Segment
Application
Microorganism Suitability
Sterility
Sterilization Method
Showing 1-1 of 1