Skip to Content
FI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,4-Dioxane
Synonym(s):
Diethylene oxide, Dioxane, 1,4-Dioxane,
p
-Dioxane
Linear Formula:
-O(CH
2
)
2
O(CH
2
)
2
-
CAS Number:
123-91-1
Molecular Weight:
88.11
EC Number:
204-661-8
Products
Product Category
solvents (8)
Brand
Sigma-Aldrich (5)
Supelco (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (2)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
360481
1,4-Dioxane,
ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Expand
View Pricing
5.89591
1,4-Dioxane,
anhydrous, 99.8%
Expand
View Pricing
34857
1,4-Dioxane,
suitable for HPLC, ≥99.5%
Expand
View Pricing
676934
1,4-Dioxane,
ACS reagent, ≥99.0%
Expand
View Pricing
1.09671
1,4-Dioxane,
for analysis EMSURE
®
ACS,ISO
Expand
View Pricing
1.03115
1,4-Dioxane,
EMPLURA
®
Expand
View Pricing
1.03132
1,4-Dioxane,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
DX2090
1,4-Dioxane,
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
900640
1,4-Dioxane,
anhydrous, ZerO
2
®
, 99.8%
Expand
View Pricing