Skip to Content
Merck
Loperamide Hydrochloride

Loperamide hydrochloride

Loperamide Hydrochloride
Synonym(s):
4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride, Loperamide Hydrochloride
Empirical Formula (Hill Notation):
C29H33ClN2O2 · xHCl
CAS Number:
Molecular Weight:
477.04 (free base basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing