Trypsin inhibitor from Glycine max (soybean)
Synonym(s):
SBTI, Ovomucoid, Kunitz Inhibitor, Trypsin Inhibitor from lima beans, Trypsin Inhibitor, Soybean, Trypsin inhibitor from Glycine max (soybean), Trypsin Inhibitor, Soybean, High Activity
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Product Line
Research Area
Tissue
Quality Segment
Application
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing