Carboxymethylcellulose sodium salt
Synonym(s):
Sodium carboxymethylcellulose, Carboxymethylcellulose sodium salt, Carboxymethylcellulose, Sodium Salt, Low Viscosity, Carboxymethylcellulose, Sodium Salt, High Viscosity, Aquacide II
CAS Number:
Products
Product Category
Brand
Color
Feature
Melting Point (°C)
pH Value
Physical Form
Agency
Tissue
Quality Segment
Polymer Composition
Polymer Type
Showing 1-14 of 14
Compare
Product Number
Description
Pricing