Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1571

Sigma-Aldrich

Anti-ARHGEF9 Antibody, clone 4D7.1

clone 4D7.1, from mouse

Synonym(s):

Rho guanine nucleotide exchange factor 9, Collybistin, PEM-2 homolog, Rac/Cdc42 guanine nucleotide exchange factor 9

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D7.1, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

General description

Rho guanine nucleotide exchange factor 9 (UniProt O43307; also known as Collybistin, PEM-2 homolog, Rac/Cdc42 guanine nucleotide exchange factor 9) is encoded by the ARHGEF9 (also known as ARHDH9, EIEE8, KIAA0424) gene (Gene ID 23229) in human. Guanine nucleotide exchange factors (GEFs) are cytoplasmic proteins that activate GTPases by stimulating the release of guanosine diphosphate (GDP) to allow binding of guanosine triphosphate (GTP). ARHGEF9 acts as a specific GEF for CDC42 and promotes formation of GPHN clusters. In the brain tissue it interacts with the receptor-anchoring protein gephyrin to activate dc42, which is known to regulate actin cytoskeleton dynamics. Its binding to gephyrin can also modulate GABAergic and glycinergic neurotransmission at the synapse. Abnormalities in its expression have been linked to epileptic encephalopathy, early infantile, 8.

Specificity

Clone 4D7.1 targets ARHGEF9 C-terminal region present in human isoforms 1/2/3 and mouse isoforms 1/2/4 reported by UniProt (O43307 and Q3UTH8). Reactivity toward mouse isoform 3 is not expected.

Immunogen

GST-tagged human ARHGEF9 C-terminal fragment.

Application

Anti-ARHGEF9 Antibody, clone 4D7.1, Cat. No. MABN1571, is a highly specific mouse monoclonal antibody that targets ARHGEF9 and has been tested in Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected ARHGEF9 in 10 µg of mouse whole brain tissue lysate.

Quality

Evaluated by Western Blotting in mouse hypothalamus tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ARHGEF9 in 10 µg of mouse hypothalamus tissue lysate.

Target description

~61 kDa observed. 60.98/54.97/49.46 kDa (human isoform 1/2/3) and 60.93/58.62/60.86 kDa (mouse isoform 1/2/4) calculated. Human isoforms 1 and 2 are also known as CB3(SH3+) and CB3(SH3-). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service